Assay Details
| Gene Symbol: MIR2394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031170.1 | 1 | 24 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | survival motor neuron domain containing 1 | SMNDC1 | SPF30 | -171.7kb | Sense | Chr. 26 - 30879035 - 30890603 on NCBI Build ARS-UCD1.2 |
| Downstream | dual specificity phosphatase 5 | DUSP5 | 1.3kb | Anti-sense | Chr. 26 - 31063686 - 31076459 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2394
Genomic Map