Assay Details
| Gene Symbol: MIR2349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031281.1 | 1 | 28 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TIMP metallopeptidase inhibitor 2 | TIMP2 | -90.7kb | Anti-sense | Chr. 19 - 53460835 - 53512593 on NCBI Build ARS-UCD1.2 | |
| Downstream | phosphatidylglycerophosphate synthase 1 | PGS1 | 153.7kb | Sense | Chr. 19 - 53757025 - 53824460 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2349
Genomic Map