Assay Details
| Gene Symbol: MIR2346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031285.1 | 1 | 44 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | centromere protein X | CENPX |
BHLHB2, STRA13 |
-15.8kb | Anti-sense | Chr. 19 - 50887829 - 50891455 on NCBI Build ARS-UCD1.2 |
| Downstream | myeloid associated differentiation marker like 2 | MYADML2 | 45.7kb | Anti-sense | Chr. 19 - 50953006 - 50956142 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2346
Genomic Map