Assay Details
| Gene Symbol: MIR1301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030823.1 | 1 | 41 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DNA methyltransferase 3 alpha | DNMT3A | Sense | Chr. 11 - 73971560 - 74074893 on NCBI Build ARS-UCD1.2 | ||
| Downstream | proopiomelanocortin | POMC | 132.1kb | Sense | Chr. 11 - 74117368 - 74124990 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR1301
Genomic Map