Assay Details
| Gene Symbol: MIR2413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031309.1 | 1 | 39 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | colony stimulating factor 1 | CSF1 | -79.0kb | Sense | Chr. 3 - 33489270 - 33509364 on NCBI Build ARS-UCD1.2 | |
| Downstream | EPS8 like 3 | EPS8L3 | 46.3kb | Anti-sense | Chr. 3 - 33634756 - 33649761 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2413
Genomic Map