Assay Details
| Gene Symbol: MIR374B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031314.1 | 1 | 46 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-545 | MIR545 |
MIRN545, bta-mir-545 |
-72.1kb | Sense | Chr. X - 76853198 - 76853303 on NCBI Build ARS-UCD1.2 |
| Downstream | microRNA mir-421 | MIR421 |
bta-mir-421, mir-421 |
74.0 | Sense | Chr. X - 76925575 - 76925659 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR374B
Genomic Map