Assay Details
| Gene Symbol: MIR2411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030848.1 | 1 | 37 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin associated protein 2 like | UBAP2L | Sense | Chr. 3 - 16253211 - 16301183 on NCBI Build ARS-UCD1.2 | ||
| Downstream | chromosome 3 C1orf43 homolog | C3H1orf43 | C1orf43 | 30.9kb | Anti-sense | Chr. 3 - 16300660 - 16309171 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2411
Genomic Map