Assay Details
| Gene Symbol: MIR2332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030857.1 | 1 | 36 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UTP6 small subunit processome component | UTP6 | Sense | Chr. 19 - 18221985 - 18249179 on NCBI Build ARS-UCD1.2 | ||
| Downstream | coordinator of PRMT5 and differentiation stimulator | COPRS |
C19H17orf79, COPR5 |
22.6kb | Sense | Chr. 19 - 18255752 - 18260611 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2332
Genomic Map