Assay Details
| Gene Symbol: MIR584-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031334.1 | 1 | 49 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane BAX inhibitor motif containing 7 | TMBIM7 | TMBIM1B | Sense | Chr. 4 - 9735082 - 9759260 on NCBI Build ARS-UCD1.2 | |
| Downstream | GATA zinc finger domain containing 1 | GATAD1 | 27.5kb | Anti-sense | Chr. 4 - 9764481 - 9777766 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR584-5
Genomic Map