Assay Details
| Gene Symbol: MIR545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031358.1 | 1 | 55 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-374a | MIR374A |
MIRN374, MIRN374A, bta-mir-374, bta-mir-374a, mir-374a |
-79.0 | Sense | Chr. X - 76853048 - 76853119 on NCBI Build ARS-UCD1.2 |
| Downstream | microRNA mir-374b | MIR374B |
bta-mir-374b, mir-374b |
72.1kb | Sense | Chr. X - 76925407 - 76925501 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR545
Genomic Map