Assay Details
| Gene Symbol: MIR2328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031389.1 | 1 | 40 | 52 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | IST1 factor associated with ESCRT-III | IST1 | -12.0kb | Anti-sense | Chr. 18 - 39158473 - 39203967 on NCBI Build ARS-UCD1.2 | |
| Downstream | ataxin 1 like | ATXN1L | 18.8kb | Anti-sense | Chr. 18 - 39234800 - 39245938 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2328
Genomic Map