Assay Details
| Gene Symbol: MIR2891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036358.1 | 1 | 38 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial calcium uptake 1 | MICU1 | CBARA1 | Anti-sense | Chr. 28 - 28402870 - 28619032 on NCBI Build ARS-UCD1.2 | |
| Downstream | mitochondrial calcium uniporter | MCU | CCDC109A | 216.0kb | Sense | Chr. 28 - 28709011 - 28904117 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2891
Genomic Map