Assay Details
| Gene Symbol: LOC100196901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038105.2 | 2 - 3 | 173 | 66 | ||
| NR_038106.2 | 2 - 3 | 173 | 66 | |||
| NR_038107.2 | 2 - 3 | 173 | 66 | |||
| GenBank mRNA | FJ357710.1 | - | 2 - 3 | 151 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2 | ST6GALNAC2 | -3.3kb | Anti-sense | Chr. 19 - 55174159 - 55194286 on NCBI Build ARS-UCD1.2 | |
| Downstream | cytoglobin | CYGB | GLNF | 19.9kb | Anti-sense | Chr. 19 - 55218139 - 55227970 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

LOC100196901
Genomic Map