Assay Details
| Gene Symbol: LOC100848703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045646.1 | 1 - 2 | 1118 | 138 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB33B, member RAS oncogene family |
RAB33B | -14.0 | Anti-sense | Chr. 17 - 18245542 - 18265684 on NCBI Build ARS-UCD1.2 | |
| Downstream |
N-alpha-acetyltransferase 15, NatA auxiliary subunit |
NAA15 | NARG1 | 37.4kb | Anti-sense | Chr. 17 - 18304431 - 18377855 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

LOC100848703
Genomic Map