Assay Details
| Gene Symbol: MIR6522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_107762.1 | 1 | 34 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | A-kinase anchoring protein 6 | AKAP6 | -177.1kb | Sense | Chr. 21 - 42714636 - 43215346 on NCBI Build ARS-UCD1.2 | |
| Downstream | egl-9 family hypoxia inducible factor 3 | EGLN3 | 994.6kb | Anti-sense | Chr. 21 - 44387137 - 44416154 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR6522
Genomic Map