Assay Details
| Gene Symbol: MIR6528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_107797.1 | 1 | 36 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-670 | MIR670 | bta-mir-670 | -24.5kb | Sense | Chr. 15 - 73648217 - 73648315 on NCBI Build ARS-UCD1.2 |
| Downstream | microRNA mir-129-2 | MIR129-2 |
bta-mir-129-2, mir-129-2 |
1.2kb | Sense | Chr. 15 - 73674100 - 73674193 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR6528
Genomic Map