Assay Details
| Gene Symbol: KANTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132122.1 | 2 - 3 | 170 | 85 | ||
| GenBank mRNA | BM258305.1 | - | 2 - 3 | 135 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor 173 | GPR173 | -13.5kb | Sense | Chr. X - 90534206 - 90558458 on NCBI Build ARS-UCD1.2 | |
| Downstream | lysine demethylase 5C | KDM5C |
JARID1C, SMCX |
49.0kb | Anti-sense | Chr. X - 90644122 - 90675153 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

KANTR
Genomic Map