Assay Details
Gene Symbol: MTLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_132744.1 | 1 - 2 | 333 | 100 | ||
GenBank mRNA | BM258536.1 | - | 1 - 2 | 307 | 100 | |
CN439569.1 | - | 1 - 2 | 210 | 100 | ||
EV610749.1 | - | 1 - 2 | 297 | 100 | ||
EV783903.1 | - | 1 - 2 | 114 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | two pore channel 3 | TPC3 | -17.3kb | Anti-sense | Chr. 11 - 1621153 - 1675048 on NCBI Build ARS-UCD1.2 | |
Downstream | nephrocystin 1 | NPHP1 | 3.2kb | Sense | Chr. 11 - 1696581 - 1763883 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
