Assay Details
| Gene Symbol: AIFM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314031.1 | NP_001300960.1 | 8 - 9 | 1000 | 60 | |
| GenBank mRNA | BC142171.1 | - | 8 - 9 | 1000 | 60 | |
More Information
Set Membership: |
|

AIFM2
Genomic Map