Assay Details
| Gene Symbol: CD1B5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193297.1 | NP_001180226.1 | 2 - 3 | 434 | 86 | |
| GenBank mRNA | EH148414.1 | - | 2 - 3 | 435 | 86 | |
| EH160235.1 | - | 2 - 3 | 345 | 86 | ||
| GU325785.1 | - | 2 - 3 | 348 | 86 | ||
| JN033695.1 | - | 2 - 3 | 329 | 86 | ||
More Information
Set Membership: |
|

CD1B5
Genomic Map