Assay Details
| Gene Symbol: IFNL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281901.1 | NP_001268830.1 | 2 - 3 | 268 | 108 | |
| GenBank mRNA | HQ317919.1 | - | 2 - 3 | 268 | 108 | |
More Information
Set Membership: |
|

IFNL3
Genomic Map