Assay Details
| Gene Symbol: OVOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302760.1 | NP_001289689.1 | 2 - 3 | 158 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OVOL3| Gene Symbol: OVOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302760.1 | NP_001289689.1 | 2 - 3 | 158 | 99 | |
Set Membership: |
|