Assay Details
| Gene Symbol: DUSP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304282.2 | NP_001291211.1 | 1 - 2 | 543 | 102 | |
| GenBank mRNA | CR852509.2 | - | 1 - 2 | 304 | 102 | |
More Information
Set Membership: |
|

DUSP5
Genomic Map