Assay Details
| Gene Symbol: DUSP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304282.2 | NP_001291211.1 | 2 - 3 | 696 | 87 | |
| GenBank mRNA | CR852509.2 | - | 2 - 3 | 457 | 87 | |
More Information
Set Membership: |
|

DUSP5
Genomic Map