Assay Details
| Gene Symbol: NTF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304828.1 | NP_001291757.1 | 2 - 3 | 256 | 88 | |
| GenBank mRNA | EH210175.1 | - | 2 - 3 | 257 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001304828.1 transcripts.
Set Membership: |
|

NTF4
Genomic Map