Assay Details
| Gene Symbol: APELA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308249.1 | NP_001295178.1 | 1 - 2 | 273 | 136 | |
| GenBank mRNA | CK832483.1 | - | 1 - 2 | 201 | 136 | |
| EV797665.1 | - | 1 - 2 | 278 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001308249.1 transcripts.
Set Membership: |
|

APELA
Genomic Map