Assay Details
| Gene Symbol: RACK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175802.3 | NP_786996.1 | 7 - 8 | 1009 | 67 | |
| GenBank mRNA | BC102286.1 | - | 7 - 8 | 990 | 67 | |
| EE233795.1 | - | 3 - 4 | 375 | 67 | ||
| EV698190.1 | - | 1 - 2 | 145 | 67 | ||
| EW687929.1 | - | 1 - 2 | 145 | 67 | ||
More Information
Set Membership: |
|

RACK1
Genomic Map