Assay Details
| Gene Symbol: ZNF354A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256565.1 | NP_001243494.1 | 1 - 2 | 166 | 123 | |
| GenBank mRNA | EH150832.1 | - | 1 - 2 | 167 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001256565.1 transcripts.
Set Membership: |
|

ZNF354A
Genomic Map