Assay Details
Gene Symbol: ZNF354A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256565.1 | NP_001243494.1 | 3 - 4 | 376 | 85 | |
GenBank mRNA | EH150832.1 | - | 3 - 4 | 377 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
