Assay Details
| Gene Symbol: LOC104968411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC149909.1 | - | 10 - 11 | 1524 | 122 | |
| U63924.1 | - | 47 - 48 | 7274 | 122 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC104968411| Gene Symbol: LOC104968411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC149909.1 | - | 10 - 11 | 1524 | 122 | |
| U63924.1 | - | 47 - 48 | 7274 | 122 | ||
Set Membership: |
|