Assay Details
| Gene Symbol: ERICH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC126653.1 | - | 9 - 10 | 1312 | 89 | |
| BC142436.1 | - | 4 - 5 | 764 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH3| Gene Symbol: ERICH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC126653.1 | - | 9 - 10 | 1312 | 89 | |
| BC142436.1 | - | 4 - 5 | 764 | 89 | ||
Set Membership: |
|