Assay Details
| Gene Symbol: IGHM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF005274.1 | - | 3 - 4 | 970 | 97 | |
| AF097213.1 | - | 3 - 4 | 979 | 97 | ||
| AY221099.1 | - | 3 - 4 | 953 | 97 | ||
| BC114850.1 | - | 5 - 6 | 1433 | 97 | ||
| BC147886.1 | - | 6 - 7 | 1461 | 97 | ||
More Information
Set Membership: |
|

IGHM
Genomic Map