Assay Details
| Gene Symbol: COASY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC109707.1 | - | 1 - 2 | 91 | 60 | |
| BC146136.1 | - | 1 - 2 | 96 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

COASY| Gene Symbol: COASY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC109707.1 | - | 1 - 2 | 91 | 60 | |
| BC146136.1 | - | 1 - 2 | 96 | 60 | ||
Set Membership: |
|