Assay Details
| Gene Symbol: BEND7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001369852.1 | NP_001356781.1 | 3 - 4 | 832 | 91 | |
| GenBank mRNA | BC111251.1 | - | 4 - 5 | 694 | 91 | |
More Information
Set Membership: |
|

BEND7
Genomic Map