Assay Details
| Gene Symbol: nlp-40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306277.3 | NP_001293206.1 | 1 - 2 | 57 | 61 | |
| NM_058259.5 | NP_490661.1 | 3 - 4 | 348 | 61 | ||
More Information
Set Membership: |
|

nlp-40
Genomic Map