Assay Details
| Gene Symbol: rcor-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306278.3 | NP_001293207.1 | 1 - 2 | 57 | 69 | |
| NM_058261.5 | NP_490662.1 | 2 - 3 | 153 | 69 | ||
| NM_058262.6 | NP_490663.1 | 2 - 3 | 144 | 69 | ||
More Information
Set Membership: |
|

rcor-1
Genomic Map