Assay Details
| Gene Symbol: pgs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058265.6 | NP_490666.2 | 1 - 2 | 89 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pgs-1| Gene Symbol: pgs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058265.6 | NP_490666.2 | 1 - 2 | 89 | 56 | |
Set Membership: |
|