Assay Details
| Gene Symbol: pgs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377579.2 | NP_001364825.1 | 3 - 4 | 223 | 71 | |
| NM_058265.6 | NP_490666.2 | 5 - 6 | 622 | 71 | ||
More Information
Set Membership: |
|

pgs-1
Genomic Map