Assay Details
| Gene Symbol: slt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078036.7 | NP_510437.2 | 24 - 25 | 3293 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

slt-1| Gene Symbol: slt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078036.7 | NP_510437.2 | 24 - 25 | 3293 | 107 | |
Set Membership: |
|