Assay Details
| Gene Symbol: dpf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078060.10 | NP_510461.1 | 13 - 14 | 2102 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpf-2| Gene Symbol: dpf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078060.10 | NP_510461.1 | 13 - 14 | 2102 | 118 | |
Set Membership: |
|