Assay Details
| Gene Symbol: dpf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078060.10 | NP_510461.1 | 16 - 17 | 2412 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpf-2| Gene Symbol: dpf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078060.10 | NP_510461.1 | 16 - 17 | 2412 | 79 | |
Set Membership: |
|