Assay Details
| Gene Symbol: mlt-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078110.4 | NP_510511.1 | 8 - 9 | 1304 | 69 | |
| NM_078111.4 | NP_510512.1 | 8 - 9 | 1246 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mlt-9| Gene Symbol: mlt-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078110.4 | NP_510511.1 | 8 - 9 | 1304 | 69 | |
| NM_078111.4 | NP_510512.1 | 8 - 9 | 1246 | 69 | ||
Set Membership: |
|