Assay Details
| Gene Symbol: ocr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078119.7 | NP_510520.4 | 1 - 2 | 166 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ocr-3| Gene Symbol: ocr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078119.7 | NP_510520.4 | 1 - 2 | 166 | 93 | |
Set Membership: |
|