Assay Details
| Gene Symbol: kin-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029498.6 | NP_001024669.1 | 1 - 2 | 592 | 59 | |
| NM_001373594.3 | NP_001359567.1 | 2 | 642 | 59 | ||
| NM_001381166.1 | NP_001367127.1 | 1 - 2 | 592 | 59 | ||
| GenBank mRNA | AY836556.1 | - | 1 - 2 | 557 | 59 | |
| AY836558.1 | - | 1 - 2 | 642 | 59 | ||
More Information
Set Membership: |
|

kin-20
Genomic Map