Assay Details
| Gene Symbol: kin-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029498.6 | NP_001024669.1 | 2 - 3 | 1023 | 146 | |
| NM_001373594.3 | NP_001359567.1 | 3 | 1073 | 146 | ||
| NM_001373595.3 | NP_001360748.1 | 2 - 3 | 500 | 146 | ||
| NM_001375178.1 | NP_001362050.1 | 1 - 2 | 430 | 146 | ||
| NM_001381166.1 | NP_001367127.1 | 2 - 3 | 1023 | 146 | ||
| NM_001383681.1 | 1 - 2 | 430 | 146 | |||
| NM_182447.10 | NP_872247.4 | 1 - 2 | 435 | 146 | ||
| GenBank mRNA | AY836556.1 | - | 2 - 3 | 988 | 146 | |
| AY836557.1 | - | 3 - 4 | 530 | 146 | ||
| AY836558.1 | - | 2 - 3 | 1073 | 146 | ||
More Information
Set Membership: |
|

kin-20
Genomic Map