Assay Details
| Gene Symbol: suox-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029564.6 | NP_001024735.1 | 3 - 4 | 491 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

suox-1| Gene Symbol: suox-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029564.6 | NP_001024735.1 | 3 - 4 | 491 | 115 | |
Set Membership: |
|