Assay Details
Gene Symbol: acox-1.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078202.9 | NP_510603.2 | 1 - 2 | 138 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
