Assay Details
| Gene Symbol: acox-1.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078202.9 | NP_510603.2 | 3 - 4 | 388 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acox-1.6| Gene Symbol: acox-1.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078202.9 | NP_510603.2 | 3 - 4 | 388 | 83 | |
Set Membership: |
|