Assay Details
| Gene Symbol: mrp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373606.3 | NP_001359552.1 | 19 - 20 | 3742 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrp-3| Gene Symbol: mrp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373606.3 | NP_001359552.1 | 19 - 20 | 3742 | 122 | |
Set Membership: |
|