Assay Details
| Gene Symbol: F09C8.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078235.7 | NP_510636.2 | 7 - 8 | 809 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F09C8.1| Gene Symbol: F09C8.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078235.7 | NP_510636.2 | 7 - 8 | 809 | 93 | |
Set Membership: |
|